Cyclopentylboronsäure 97%
|
|
BLDPBD2445-1GEA
10
EUR
BLDPBD2445-1G
Cyclopentylboronsäure 97%
Cyclopentylboronic acid
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₅H₁₁BO₂ Molekulargewicht: 113,95 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD01074541 CAS-Nummer: 63076-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...