6-Methoxy-5-nitro (1H)indazole 95%
|
|
BLDPBD245184-100MGEA
59
EUR
BLDPBD245184-100MG
BLDPBD245184-1G
BLDPBD245184-5G
BLDPBD245184-250MG
6-Methoxy-5-nitro (1H)indazole 95%
6-Methoxy-5-nitro (1H)indazole
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₇N₃O₃ Molekulargewicht: 193,159 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11007881 CAS-Nummer: 152626-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...