4-Brom-3-chlor-2-nitroanilin 95%

BLDPBD245823-100MGEA 45 EUR
BLDPBD245823-100MG BLDPBD245823-1G BLDPBD245823-250MG
4-Brom-3-chlor-2-nitroanilin 95%
4-Brom-3-chlor-2-nitroanilin

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₄BrClN₂O₂
Molekulargewicht: 251,4652 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09878187
CAS-Nummer: 1000573-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR