(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on 98%

BLDPBD246816-10GEA 11 EUR
BLDPBD246816-10G BLDPBD246816-25G BLDPBD246816-100G BLDPBD246816-500G BLDPBD246816-5G BLDPBD246816-0.5G
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on 98%
(±)-Naringenin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₅H₁₂O₅
Molekulargewicht: 272,25 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006844
CAS-Nummer: 67604-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR