(R)-(-)-3-Aminopiperidin Dihydrochlorid 97%

BLDPBD2500-500GEA 696 EUR
BLDPBD2500-500G BLDPBD2500-1G BLDPBD2500-100G BLDPBD2500-5G BLDPBD2500-25G
(R)-(-)-3-Aminopiperidin Dihydrochlorid 97%
(R)-(-)-3-Aminopiperidin Dihydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₄Cl₂N₂
Molekulargewicht: 173,08 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06799458
CAS-Nummer: 334618-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR