2-Amino-4-methoxybenzonitril 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD25087-250MGEA 16 EUR
BLDPBD25087-250MG BLDPBD25087-10G BLDPBD25087-1G BLDPBD25087-25G BLDPBD25087-5G
2-Amino-4-methoxybenzonitril 97%
2-Amino-4-methoxybenzonitril

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈N₂O
Molekulargewicht: 148,1600 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05149246
CAS-Nummer: 38487-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR