4-Bromo-2,6-dimethoxybenzaldehyde 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD255755-1GEA 24 EUR
BLDPBD255755-1G BLDPBD255755-250MG BLDPBD255755-5G BLDPBD255755-10G BLDPBD255755-100G BLDPBD255755-25G BLDPBD255755-100MG
4-Bromo-2,6-dimethoxybenzaldehyde 97%
4-Bromo-2,6-dimethoxybenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉BrO₃
Molekulargewicht: 245,07 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22192498
CAS-Nummer: 1354050-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR