4'-Fluor-3'-hydroxyacetophenon 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD257961-1GEA 121 EUR
BLDPBD257961-1G BLDPBD257961-250MG BLDPBD257961-5G BLDPBD257961-100MG
4'-Fluor-3'-hydroxyacetophenon 98%
4'-Fluor-3'-hydroxyacetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇FO₂
Molekulargewicht: 154,14 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12545938
CAS-Nummer: 949159-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR