Methyl-2-brom-1,3-thiazol-4-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD2585-25GEA 148 EUR
BLDPBD2585-25G BLDPBD2585-5G BLDPBD2585-100G BLDPBD2585-1G BLDPBD2585-250MG BLDPBD2585-10G
Methyl-2-brom-1,3-thiazol-4-carboxylat 97%
Methyl-2-bromothiazole-4-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₄BrNO₂S
Molekulargewicht: 222,06 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659908
CAS-Nummer: 170235-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR