4-(4'-Fluorbenzyloxy)phenylboronsäure 95%

BLDPBD261037-25GEA 442 EUR
BLDPBD261037-25G BLDPBD261037-5G BLDPBD261037-250MG BLDPBD261037-1G
4-(4'-Fluorbenzyloxy)phenylboronsäure 95%
4-(4'-Fluorbenzyloxy)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₂BFO₃
Molekulargewicht: 246,0420 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07784403
CAS-Nummer: 871125-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR