3-Aminopiperidine hydrochloride, N1-CBZ protected 97%

BLDPBD261107-5GEA 159 EUR
BLDPBD261107-5G BLDPBD261107-10G BLDPBD261107-25G BLDPBD261107-1G BLDPBD261107-250MG
3-Aminopiperidine hydrochloride, N1-CBZ protected 97%
3-Aminopiperidine hydrochloride, N1-CBZ protected

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₈N₂O₂·ClH
Molekulargewicht: 270,755 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD04038445
CAS-Nummer: 960541-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR