5-Brom-2-chlorbenzaldehyd 98%

BLDPBD26181-10GEA 15 EUR
BLDPBD26181-10G BLDPBD26181-500G BLDPBD26181-100G BLDPBD26181-1G BLDPBD26181-5G BLDPBD26181-25G
5-Brom-2-chlorbenzaldehyd 98%
5-Brom-2-chlorbenzaldehyd

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₄BrClO
Molekulargewicht: 219,46 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08445659
CAS-Nummer: 189628-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR