3',5'-Diethoxyacetophenon 98%

BLDPBD261986-25GEA 380 EUR
BLDPBD261986-25G BLDPBD261986-1G BLDPBD261986-0.025 BLDPBD261986-5G
3',5'-Diethoxyacetophenon 98%
3',5'-Diethoxyacetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₃
Molekulargewicht: 208,2536 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02258864
CAS-Nummer: 103604-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR