5-Fluor-2-methoxy-4-pyridinboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD26285-1GEA 77 EUR
BLDPBD26285-1G BLDPBD26285-100MG
5-Fluor-2-methoxy-4-pyridinboronsäure 95%
5-Fluoro-2-methoxy-4-pyridineboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₆H₇BFNO₃
Molekulargewicht: 170,9300 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD06657887
CAS-Nummer: 1043869-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR