2-Amino-5-fluor-3-methylpyridin 98%

BLDPBD26295-250MGEA 33 EUR
BLDPBD26295-250MG BLDPBD26295-100MG
2-Amino-5-fluor-3-methylpyridin 98%
2-Amino-5-fluoro-3-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₇FN₂
Molekulargewicht: 126,1300 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07375130
CAS-Nummer: 886365-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR