3-Methoxy-2-naphthalinboronsäure 98%

BLDPBD263160-250MGEA 6 EUR
BLDPBD263160-250MG BLDPBD263160-5G BLDPBD263160-25G BLDPBD263160-0.025 BLDPBD263160-10G BLDPBD263160-1G BLDPBD263160-100G
3-Methoxy-2-naphthalinboronsäure 98%
3-Methoxy-2-naphthaleneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₁BO₃
Molekulargewicht: 202,0100 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD12025968
CAS-Nummer: 104115-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR