4-Chlor-3,5-difluorbenzaldehyd 98+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD265712-1GEA 82 EUR
BLDPBD265712-1G BLDPBD265712-250MG BLDPBD265712-5G BLDPBD265712-100MG
4-Chlor-3,5-difluorbenzaldehyd 98+%
4-Chloro-3,5-difluorobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₃ClF₂O
Molekulargewicht: 176,5479 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11553555
CAS-Nummer: 1160573-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR