5-Bromofuran-2-carbonitrile 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD266644-250MGEA 34 EUR
BLDPBD266644-250MG BLDPBD266644-5G BLDPBD266644-1G BLDPBD266644-100MG
5-Bromofuran-2-carbonitrile 97%
5-Bromofuran-2-carbonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₂BrNO
Molekulargewicht: 171,979 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03410206
CAS-Nummer: 4915-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR