2-Methylbenzo[b]thiophene-3-carboxaldehyde 95%

BLDPBD276463-1GEA 133 EUR
BLDPBD276463-1G BLDPBD276463-100MG BLDPBD276463-5G BLDPBD276463-250MG
2-Methylbenzo[b]thiophene-3-carboxaldehyde 95%
2-Methylbenzo[b]thiophene-3-carboxaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈OS
Molekulargewicht: 176,235 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01651753
CAS-Nummer: 30446-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR