Ethyl-4-oxazolcarboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD28118-250MGEA 6 EUR
BLDPBD28118-250MG BLDPBD28118-1G BLDPBD28118-25G BLDPBD28118-100G BLDPBD28118-10G BLDPBD28118-5G
Ethyl-4-oxazolcarboxylat 97%
Ethyl-4-oxazolcarboxylat

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₇NO₃
Molekulargewicht: 141,1300 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04114940
CAS-Nummer: 23012-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR