Pyrazolo[1,5-a]pyridin 97%

BLDPBD28243-250MGEA 50 EUR
BLDPBD28243-250MG BLDPBD28243-25G BLDPBD28243-1G BLDPBD28243-10G BLDPBD28243-5G BLDPBD28243-100MG
Pyrazolo[1,5-a]pyridin 97%
Pyrazolo[1,5-a]pyridin

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₆N₂
Molekulargewicht: 118,14 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08752622
CAS-Nummer: 274-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR