Methyl-4-brompicolinat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD28303-500GEA 1995 EUR
BLDPBD28303-500G BLDPBD28303-25G BLDPBD28303-5G BLDPBD28303-1G BLDPBD28303-250MG BLDPBD28303-100G BLDPBD28303-10G
Methyl-4-brompicolinat 97%
Methyl-4-brompicolinat

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BrNO₂
Molekulargewicht: 216,03 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07374897
CAS-Nummer: 29681-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR