3-Iodimidazo[1,2-a]pyridin 95%
|
|
BLDPBD28328-250MGEA
45
EUR
BLDPBD28328-250MG
BLDPBD28328-5G
BLDPBD28328-1G
BLDPBD28328-10G
BLDPBD28328-25G
3-Iodimidazo[1,2-a]pyridin 95%
3-Iodimidazo[1,2-a]pyridin
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₇H₅IN₂ Molekulargewicht: 244,0300 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD06739222 CAS-Nummer: 307503-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...