3-Isoxazolcarbonsäure 96%

BLDPBD28350-10GEA 110 EUR
BLDPBD28350-10G BLDPBD28350-5G BLDPBD28350-250MG BLDPBD28350-1G BLDPBD28350-100MG BLDPBD28350-25G
3-Isoxazolcarbonsäure 96%
3-Isoxazolecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄H₃NO₃
Molekulargewicht: 113,07 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06738796
CAS-Nummer: 3209-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR