Methyl 3-bromo-2-(bromomethyl)benzoate 95%

BLDPBD28414-5GEA 228 EUR
BLDPBD28414-5G BLDPBD28414-25G BLDPBD28414-250MG BLDPBD28414-1G BLDPBD28414-100MG
Methyl 3-bromo-2-(bromomethyl)benzoate 95%
Methyl 3-bromo-2-(bromomethyl)benzoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈Br₂O₂
Molekulargewicht: 307,967 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06804569
CAS-Nummer: 337536-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR