β-Jonon 95%

BLDPBD284140-25GEA 7 EUR
BLDPBD284140-25G BLDPBD284140-500G BLDPBD284140-100G BLDPBD284140-1000G
β-Jonon 95%
β-Jonon

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₂₀O
Molekulargewicht: 192,30 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00001549
CAS-Nummer: 79-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR