Methyl-1H-indazol-5-carboxylat 98%
|
|
BLDPBD28804-10GEA
50
EUR
BLDPBD28804-10G
BLDPBD28804-250MG
BLDPBD28804-500G
BLDPBD28804-100G
BLDPBD28804-1G
BLDPBD28804-25G
BLDPBD28804-5G
Methyl-1H-indazol-5-carboxylat 98%
Methyl-1H-indazol-5-carboxylat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₈N₂O₂ Molekulargewicht: 176,1700 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07371609 CAS-Nummer: 473416-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...