4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD290394-250MGEA 57 EUR
BLDPBD290394-250MG BLDPBD290394-1G BLDPBD290394-100MG
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile 97%
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆ClN₃S
Molekulargewicht: 199,661 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD28134480
CAS-Nummer: 1208170-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR