Fmoc-NH-PEG3-CH2COOH 95%

BLDPBD290447-25GEA 1634 EUR
BLDPBD290447-25G BLDPBD290447-100MG BLDPBD290447-250MG BLDPBD290447-5G BLDPBD290447-1G BLDPBD290447-10G
Fmoc-NH-PEG3-CH2COOH 95%
Fmoc-NH-PEG3-CH2COOH

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₃H₂₇NO₇
Molekulargewicht: 429,463 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD13182400
CAS-Nummer: 139338-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR