2-Chlor-4,5-difluoranilin 97%
|
|
BLDPBD290832-25GEA
178
EUR
BLDPBD290832-25G
BLDPBD290832-250MG
BLDPBD290832-1G
BLDPBD290832-10G
BLDPBD290832-5G
2-Chlor-4,5-difluoranilin 97%
2-Chlor-4,5-difluoranilin
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₆H₄ClF₂N Molekulargewicht: 163,55 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08448325 CAS-Nummer: 2613-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...