4,5-Dihydrofuran-3-boronic acid, pinacol ester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD291224-50MGEA 158 EUR
BLDPBD291224-50MG
4,5-Dihydrofuran-3-boronic acid, pinacol ester 98%
4,5-Dihydrofuran-3-boronic acid, pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₁₇BO₃
Molekulargewicht: 196,051 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD13182120
CAS-Nummer: 1046812-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR