5-Chlor-3-phenyl-2,1-benzisoxazol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD29489-25GEA 82 EUR
BLDPBD29489-25G BLDPBD29489-0.025G BLDPBD29489-100G BLDPBD29489-5G BLDPBD29489-250MG BLDPBD29489-100MG BLDPBD29489-1G
5-Chlor-3-phenyl-2,1-benzisoxazol 95%
5-Chloro-3-phenyl-2,1-benzisoxazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₈ClNO
Molekulargewicht: 229,67 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014573
CAS-Nummer: 719-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR