6-Brom-1H-indazol 98%

BLDPBD29648-500GEA 645 EUR
BLDPBD29648-500G BLDPBD29648-5G BLDPBD29648-100G BLDPBD29648-25G BLDPBD29648-1G BLDPBD29648-10G
6-Brom-1H-indazol 98%
6-Bromo-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₅BrN₂
Molekulargewicht: 197,0300 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03265457
CAS-Nummer: 79762-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR