3-Amino-6-brom-2-methoxypyridin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD29839-25GEA 366 EUR
BLDPBD29839-25G BLDPBD29839-10G BLDPBD29839-1G BLDPBD29839-5G BLDPBD29839-250MG
3-Amino-6-brom-2-methoxypyridin 97%
3-Amino-6-bromo-2-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₇BrN₂O
Molekulargewicht: 203,0400 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06738312
CAS-Nummer: 89466-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR