5-Methoxy-1H-indazol 95%
|
|
BLDPBD29923-250MGEA
17
EUR
BLDPBD29923-250MG
BLDPBD29923-5G
BLDPBD29923-1G
5-Methoxy-1H-indazol 95%
5-Methoxy-1H-indazole
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₈N₂O Molekulargewicht: 148,1600 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07781657 CAS-Nummer: 94444-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...