2-Bromo-4-hydroxybenzoic acid 95%

BLDPBD305190-25GEA 1711 EUR
BLDPBD305190-25G BLDPBD305190-1G BLDPBD305190-250MG BLDPBD305190-5G BLDPBD305190-100MG
2-Bromo-4-hydroxybenzoic acid 95%
2-Bromo-4-hydroxybenzoic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BrO₃
Molekulargewicht: 217,017 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16999959
CAS-Nummer: 28547-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR