3-Brom-4-fluorbenzylamin Hydrochlorid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD307607-25GEA 229 EUR
BLDPBD307607-25G BLDPBD307607-5G BLDPBD307607-1G
3-Brom-4-fluorbenzylamin Hydrochlorid 98%
3-Brom-4-fluorbenzylamin Hydrochlorid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₈BrClFN
Molekulargewicht: 240,50 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00143094
CAS-Nummer: 202865-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR