Ethyl 4-amino-3-bromo-5-nitrobenzoate 95%

BLDPBD315891-1GEA 108 EUR
BLDPBD315891-1G BLDPBD315891-100MG BLDPBD315891-250MG
Ethyl 4-amino-3-bromo-5-nitrobenzoate 95%
Ethyl 4-amino-3-bromo-5-nitrobenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₉BrN₂O₄
Molekulargewicht: 289,083 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00044014
CAS-Nummer: 82760-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR