2-Fluor-N-methylbenzamid 95%

BLDPBD317586-250MGEA 25 EUR
BLDPBD317586-250MG BLDPBD317586-1G BLDPBD317586-5G
2-Fluor-N-methylbenzamid 95%
2-Fluor-N-methylbenzamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈FNO
Molekulargewicht: 153,16 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01214098
CAS-Nummer: 52833-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR