2-Chloro-5-methoxypyridine 98%

BLDPBD31863-1GEA 37 EUR
BLDPBD31863-1G BLDPBD31863-25G BLDPBD31863-5G BLDPBD31863-250MG BLDPBD31863-100G BLDPBD31863-10G
2-Chloro-5-methoxypyridine 98%
2-Chloro-5-methoxypyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₆ClNO
Molekulargewicht: 143,571 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06659513
CAS-Nummer: 139585-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR