2-Hydroxypyridin 98%

Synonyme: α-Pyridon2-Pyridinol2-Pyridon

BLDPBD31897-10GEA 7 EUR
BLDPBD31897-10G BLDPBD31897-0.025G BLDPBD31897-25G BLDPBD31897-5G
2-Hydroxypyridin 98%
2-Hydroxypyridin

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₅NO
Molekulargewicht: 95,10 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00462283
CAS-Nummer: 142-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR