1-(4-Bromobenzyl)-1H-pyrazole 95%

BLDPBD320866-1GEA 230 EUR
BLDPBD320866-1G BLDPBD320866-100MG BLDPBD320866-250MG
1-(4-Bromobenzyl)-1H-pyrazole 95%
1-(4-Bromobenzyl)-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉BrN₂
Molekulargewicht: 237,096 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12068403
CAS-Nummer: 1159826-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR