3-(1,3-Oxazol-5-yl)anilin 98%

BLDPBD32106-1GEA 59 EUR
BLDPBD32106-1G BLDPBD32106-5G BLDPBD32106-10G
3-(1,3-Oxazol-5-yl)anilin 98%
3-(1,3-Oxazol-5-yl)anilin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₈N₂O
Molekulargewicht: 160,1700 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00052191
CAS-Nummer: 157837-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR