3'-Brom-2'-fluoracetophenon 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD32164-1GEA 24 EUR
BLDPBD32164-1G BLDPBD32164-100G BLDPBD32164-25G BLDPBD32164-10G BLDPBD32164-250MG BLDPBD32164-5G
3'-Brom-2'-fluoracetophenon 98%
3'-Brom-2'-fluoracetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆BrFO
Molekulargewicht: 217,04 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09264507
CAS-Nummer: 161957-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR