(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%

BLDPBD32264-1GEA 56 EUR
BLDPBD32264-1G BLDPBD32264-250MG BLDPBD32264-5G BLDPBD32264-25G
(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%
(4S)-4-Benzyl-1,3-thiazolidine-2-thione

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁NS₂
Molekulargewicht: 209,331 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06658216
CAS-Nummer: 171877-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR