Tetrahydro-4H-thiopyran-4-on 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD32283-1000GEA 1064 EUR
BLDPBD32283-1000G BLDPBD32283-5G BLDPBD32283-25G BLDPBD32283-100G BLDPBD32283-10G BLDPBD32283-500G
Tetrahydro-4H-thiopyran-4-on 98%
Tetrahydro-4H-thiopyran-4-on

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₈OS
Molekulargewicht: 116,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006660
CAS-Nummer: 1072-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR