2-Amino-3-chlor-5-(trifluormethyl)pyridin 98%

BLDPBD3244-25GEA 21 EUR
BLDPBD3244-25G BLDPBD3244-5G BLDPBD3244-500G BLDPBD3244-1000G BLDPBD3244-100G
2-Amino-3-chlor-5-(trifluormethyl)pyridin 98%
2-Amino-3-chloro-5-(trifluoromethyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₄ClF₃N₂
Molekulargewicht: 196,5600 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00042154
CAS-Nummer: 79456-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR