5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester 98%

BLDPBD324892-1GEA 176 EUR
BLDPBD324892-1G BLDPBD324892-100MG BLDPBD324892-250MG
5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester 98%
5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₁H₁₆BBrO₂S
Molekulargewicht: 303,024 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD12405479
CAS-Nummer: 1111095-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR