4-Bromo-2-(methylsulphonyl)phenol 95%

BLDPBD327905-100MGEA 72 EUR
BLDPBD327905-100MG
4-Bromo-2-(methylsulphonyl)phenol 95%
4-Bromo-2-(methylsulphonyl)phenol

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₇BrO₃S
Molekulargewicht: 251,098 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28515417
CAS-Nummer: 88041-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR