4-Brom-5-chlor-2-methylanilin 97%
|
|
BLDPBD33124-100GEA
106
EUR
BLDPBD33124-100G
BLDPBD33124-1G
BLDPBD33124-25G
BLDPBD33124-5G
BLDPBD33124-500G
4-Brom-5-chlor-2-methylanilin 97%
4-Brom-5-chlor-2-methylanilin
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₇BrClN Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
30273-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...